missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL3/CCL4 Monoclonal antibody specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | CCL3/CCL4 |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 93342 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C-C motif chemokine;C-C motif chemokine 3;G0S19-1;LD78ALPHA;MIP1A;MIP-1-alpha;SCYA3 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human CCL3/MIP-1 alpha , Ala27-Ala92, Accession # P10147 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?