Learn More
CCT8 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A7C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67539-100UL
Description
The CCT8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT8. This antibody recognizes human, mouse, rat antigen. The CCT8 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| CCT8 | |
| 1A7C3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| C21orf112, CCT theta, CCT8, Cctq, D21S246, KIAA0002, PRED71, TCP 1 theta | |
| 100 μL | |
| 10694 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |