Learn More
CCT8 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A7C3, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | CCT8 |
|---|---|
| Clone | 1A7C3 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057115
|
Proteintech
CL488-67539-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18026915
|
Proteintech
CL488-67539-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CCT8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT8. This antibody recognizes human, mouse, rat antigen. The CCT8 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| CCT8 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 10694 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1A7C3 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| C21orf112, CCT theta, CCT8, Cctq, D21S246, KIAA0002, PRED71, TCP 1 theta | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.