missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CD117/c-kit |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL1656 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL |
| Formulation | PBS (pH 7.2), 40% Glycerol |
| Gene Alias | CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein |
| Host Species | Mouse |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: VGDEIRLLCTDPGFVKWTFEILDETNENKQNEWITEKAEATNTGKYTCTNKHGLSNSIYVFVRDPAKLFLVDRSLYGKEDNDTLVRCPLTDPEVTNYSLKGCQGKPLPKDLRFIPDPKAGIMIKSVKRAYHRLCLHCSVDQ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?