missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 (Monocyte / Macrophage Marker) Mouse, Clone: LPSR/2385, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 929-MSM5-P1
Additional Details : Weight : 0.01000kg
Specifications
CD14 (Monocyte / Macrophage Marker) | |
Cell Surface | |
LPSR/2385 | |
Unconjugated | |
Mouse | |
55kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
929 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human CD14 protein (apprx. aa 25-148) | |
THP-1 cells and human monocytes. Lymph nodes and tonsils. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |