missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 (Monocyte / Macrophage Marker) Mouse, Clone: LPSR/2385, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | CD14 (Monocyte / Macrophage Marker) |
---|---|
Clone | LPSR/2385 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15931188
|
EnquireBio
929-MSM5-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921188
|
EnquireBio
929-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Especificaciones
CD14 (Monocyte / Macrophage Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human CD14 protein (apprx. aa 25-148) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
55kDa |
LPSR/2385 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
929 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface | |
THP-1 cells and human monocytes. Lymph nodes and tonsils. |