missing translation for 'onlineSavingsMsg'
Learn More

CD163 Antibody (CL10652) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30525588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525588 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525588 Supplier Novus Biologicals Supplier No. NBP343871CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD163 Monoclonal antibody specifically detects CD163 in Human,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD163
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL10652
Conjugate CoraFluor 1
Formulation PBS
Gene Alias CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q86VB7, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9332
Target Species Human, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.