Learn More
CD163 Antibody (EDHu-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand:  Novus Biologicals NB110-40686
 View more versions of this product
Product Code. 18760704
  • 580.00 € / 0.10mg
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Flow (Cell Surface).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry, Immunofluorescence
Human, Pig, Bovine, Guinea Pig, Primate, Monkey