missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (EDHu-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
249.00 € - 580.00 €
Specifications
| Antigen | CD163 |
|---|---|
| Clone | EDHu-1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay, Flow (Cell Surface) 1:10-1:1000 |
| Applications | Immunofluorescence, Flow Cytometry, Immunohistochemistry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249881
|
Novus Biologicals
NB110-40686-0.025MG |
0.025 mg |
249.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18760704
|
Novus Biologicals
NB110-40686 |
580.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Flow (Cell Surface).Specifications
| CD163 | |
| 1.0 mg/mL | |
| Immunofluorescence, Flow Cytometry, Immunohistochemistry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Sheep, Monkey, Primate, Rhesus Macaque, Guinea Pig, Human, Bovine, Pig | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| CD163 | |
| IgG1 | |
| Protein A purified | |
| NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone. |
| EDHu-1 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay, Flow (Cell Surface) 1:10-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q86VB7 | |
| 9332 | |
| Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title