missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (CB19) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Marke: Novus Biologicals NBP2-25196SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD19 Monoclonal specifically detects CD19 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro assay, CyTOF-ready.
Spezifikation
| CD19 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot reported in scientific literature (PMID 17327405), Flow Cytometry 0.2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:25-1:100, In vitro assay reported in scientific literature (PMID 9576485), CyTOF-ready | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, In vitro Assay | |
| CB19 | |
| Unconjugated | |
| P15391 | |
| CD19 | |
| Intact normal human B cell cells were used as immunogen to generate the CB19 clone. | |
| 0.025 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| Human, Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur