missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (CB19) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
187.00 € - 534.00 €
Specifications
| Antigen | CD19 |
|---|---|
| Clone | CB19 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot reported in scientific literature (PMID 17327405), Flow Cytometry 0.2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:25-1:100, In vitro assay reported in scientific literature (PMID 9576485), CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, In vitro Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18183265
|
Novus Biologicals
NBP2-25196SS |
0.025 mg |
187.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717303
|
Novus Biologicals
NBP2-25196 |
0.1 mg |
534.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal specifically detects CD19 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro assay, CyTOF-ready.Specifications
| CD19 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, In vitro Assay | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| P15391 | |
| 930 | |
| Intact normal human B cell cells were used as immunogen to generate the CB19 clone. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CB19 | |
| Western Blot reported in scientific literature (PMID 17327405), Flow Cytometry 0.2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:25-1:100, In vitro assay reported in scientific literature (PMID 9576485), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| CD19 | |
| IgG1 | |
| Protein G purified | |
| This product reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title