missing translation for 'onlineSavingsMsg'
Learn More

CD197, CCR7 Rat anti-Human, BB515, Clone: 3D12, BD Horizon™

Rat Monoclonal Antibody

Brand:  BD Biosciences 565870

 View more versions of this product

Product Code. 15884569

  • ONLINE EXCLUSIVE PRICE
    161.00 €
    155.15 € / Each
    Save 5.85 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD197, CCR7
Monoclonal
Brilliant Blue 515 (BB515)
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
25 Tests
Cell Biology
Human
Purified
Flow Cytometry
3D12
Vol. Per test: 5μL
CCR7, BLR-2, EBI-1, CMKBR7
Human CCR7 protein
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.