missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD197, CCR7 Rat anti-Human, BB515, Clone: 3D12, BD Horizon™
Rat Monoclonal Antibody
155.15 € - 331.27 €
Specifications
| Antigen | CD197, CCR7 |
|---|---|
| Clone | 3D12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15884569
|
BD Biosciences
565870 |
25 Tests |
161.00 € 155.15 € / Each Save 5.85 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15874569
|
BD Biosciences
565869 |
100 Tests |
346.00 € 331.27 € / Each Save 14.73 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.Specifications
| CD197, CCR7 | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| Rat | |
| Cell Biology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human CCR7 protein | |
| Primary | |
| Vol. Per test: 5μL |
| 3D12 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCR7, BLR-2, EBI-1, CMKBR7 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title