missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Antibody (C1A/8111R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20388-100ug
This item is not returnable.
View return policy
Description
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD1a | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Recombinant full-length human CD1a protein | |
| 100 μg | |
| Dendritic Cell Markers, Immunology | |
| 909 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| C1A/8111R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction