missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Antibody (C1A/8111R), Novus Biologicals™
Rabbit Monoclonal Antibody
298.00 € - 530.00 €
Specifications
| Antigen | CD1a |
|---|---|
| Clone | C1A/8111R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654724
|
Novus Biologicals
NBP3-20388-20ug |
20 μg |
298.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683614
|
Novus Biologicals
NBP3-20388-100ug |
100 μg |
530.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD1a | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Dendritic Cell Markers, Immunology | |
| 10mM PBS with 0.05% BSA | |
| 909 | |
| IgG κ | |
| Protein A or G purified |
| C1A/8111R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Recombinant full-length human CD1a protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title