missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a / HTA1 Rabbit, Clone: C1A/1506R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 909-RBM6-P0
This item is not returnable.
View return policy
Specifications
| CD1a / HTA1 | |
| Cell surface | |
| C1A/1506R | |
| Unconjugated | |
| NP_001754 | |
| Rabbit | |
| 49kDa | |
| Protein A/G Purified | |
| RUO | |
| 909 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| CD1A | |
| Recombinant human full-length CD1a protein | |
| MOLT-4 cells. Paracortex in a tonsil or a reactive lymph node. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction