missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a / HTA1 Rabbit, Clone: C1A/1506R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | CD1a / HTA1 |
|---|---|
| Clone | C1A/1506R |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15991148
|
EnquireBio
909-RBM6-P0 |
20 μg | |||||||
|
15901158
|
EnquireBio
909-RBM6-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD1a / HTA1 | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 909 | |
| Recombinant human full-length CD1a protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 49kDa |
| C1A/1506R | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001754 | |
| CD1A | |
| IgG κ | |
| Protein A/G Purified | |
| Cell surface | |
| MOLT-4 cells. Paracortex in a tonsil or a reactive lymph node. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title