missing translation for 'onlineSavingsMsg'
Learn More

CD20 Antibody (rMS4A1/8044), Novus Biologicals™

Product Code. p-200075933 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18610945 20 μg 20µg
18609983 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18610945 Supplier Novus Biologicals Supplier No. NBP32374420ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD20
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMS4A1/8044
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Host Species Mouse
Immunogen Recombinant fragment (around aa200-297) of human CD20 protein (exact sequence is proprietary)
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 931
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.