missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Antibody (Hu102) [Allophycocyanin], Novus Biologicals™
Shop All Bio Techne ProductsDescription
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | CD25/IL-2R alpha |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | Hu102 |
| Conjugate | APC |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formulation | PBS |
| Gene Alias | CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR |
| Host Species | Human |
| Immunogen | Human CD25/IL-2 R alpha |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?