Learn More
CD25 Rat, APC, Clone: PC61.5, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB34-APC-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15961858
  • 119.00 € / 25µg
Estimated Shipment: 05-07-2024
to see stock.
Specifications
CD25
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Il2ra
Protein A/G Purified
RUO
16184
IgG1 λ
Flow Cytometry
PC61.5
APC
NP_032393
Rat
25 μg
Primary
Store at 2-4°C.