missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Rat, APC, Clone: PC61.5, enQuireBio™
Rat Monoclonal Antibody
119.00 € - 312.00 €
Specifications
Antigen | CD25 |
---|---|
Clone | PC61.5 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961858
|
EnquireBio
QAB34-APC-25UG |
25 μg |
119.00 €
25µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951858
|
EnquireBio
QAB34-APC-100UG |
100 μg |
312.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD25 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
16184 | |
IgG1 λ | |
Protein A/G Purified |
PC61.5 | |
Flow Cytometry | |
APC | |
RUO | |
NP_032393 | |
Il2ra | |
Primary | |
Store at 2-4°C. |