missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Golden Hamster, Qfluor 630, Clone: 37.51, enQuireBio™
Golden Hamster Monoclonal Antibody
Brand: EnquireBio QAB99-QF630-500ug
This item is not returnable.
View return policy
Specifications
| CD28 | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| Cd28 | |
| Protein A/G Purified | |
| RUO | |
| 12487 | |
| IgG |
| Flow Cytometry, Immunoprecipitation | |
| 37.51 | |
| Qfluor 630 | |
| NP_031668 | |
| Golden Syrian Hamster | |
| 500 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction