missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Golden Hamster, Qfluor 630, Clone: 37.51, enQuireBio™
Golden Hamster Monoclonal Antibody
288.75 € - 420.00 €
Specifications
| Antigen | CD28 |
|---|---|
| Clone | 37.51 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15962668
|
EnquireBio
QAB99-QF630-100ug |
100 μg |
288.75 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15972668
|
EnquireBio
QAB99-QF630-500ug |
500 μg |
420.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD28 | |
| 0.5mg/mL | |
| Monoclonal | |
| Golden Syrian Hamster | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| 12487 | |
| IgG | |
| Protein A/G Purified |
| 37.51 | |
| Flow Cytometry, Immunoprecipitation | |
| Qfluor 630 | |
| RUO | |
| NP_031668 | |
| Cd28 | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title