missing translation for 'onlineSavingsMsg'
Learn More

CD30 / TNFRSF8 Mouse, Clone: CD30/412, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 943-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991208

  • 485.00 € / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD30 / TNFRSF8
Cell surface
CD30/412
Unconjugated
XP_011540745
Mouse
105-120kDa
Protein A/G Purified
RUO
943
IgG1 κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide
TNFRSF8
Human CD30 recombinant protein
Hodgkin's lymphoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers