missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 / TNFRSF8 Rabbit, Clone: Ki-1/1747R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 943-RBM7-P1
Additional Details : Weight : 0.01000kg
Specifications
CD30 / TNFRSF8 | |
Cell surface | |
Ki-1/1747R | |
Unconjugated | |
XP_011540745 | |
Rabbit | |
105-120kDa | |
Protein A/G Purified | |
RUO | |
943 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
TNFRSF8 | |
Recombinant human full-length TNFRSF8 protein | |
Hodgkin's lymphoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |