Learn More
CD31/PECAM-1 Antibody (MEC13.3) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 17 publications
Brand:  Novus Biologicals NB600-1475
 View more versions of this product
Product Code. 18737993
  • 561.00 € / 0.10mL
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD31/PECAM-1
Monoclonal
1.0 mg/mL
Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:100, In vitro assay reported in scientific literature (PMID 24647208), In vivo assay reported in scientific literature, Flow (Cell Surface) 2.5 ug/ml, CyTOF-ready
PECAM1
This CD31/PECAM-1 Antibody (MEC13.3) was developed against mouse endothelial cell line T-end.
Protein G purified
RUO
Primary
Human, Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, CyTOF
MEC13.3
Unconjugated
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, MEC 13.3 CD31, MEC 13.3 Clone, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Rat
82.5 kDa
0.1 mL
Angiogenesis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
5175
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a κ