missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31/PECAM-1 Antibody (MEC13.3) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 17 publications
190.00 € - 561.00 €
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | MEC13.3 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:100, In vitro assay reported in scientific literature (PMID 24647208), In vivo assay reported in scientific literature, Flow (Cell Surface) 2.5 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, CyTOF |
| Product Code | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18758133
|
Novus Biologicals
NB600-1475SS |
0.025 mL |
190.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18737993
|
Novus Biologicals
NB600-1475 |
0.1 mL |
561.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.Spezifikation
| CD31/PECAM-1 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, CyTOF | |
| Unconjugated | |
| Rat | |
| Angiogenesis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, MEC 13.3 CD31, MEC 13.3 Clone, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| PECAM1 | |
| IgG2a κ | |
| Protein G purified | |
| 82.5 kDa |
| MEC13.3 | |
| Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:100, In vitro assay reported in scientific literature (PMID 24647208), In vivo assay reported in scientific literature, Flow (Cell Surface) 2.5 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 5175 | |
| This CD31/PECAM-1 Antibody (MEC13.3) was developed against mouse endothelial cell line T-end. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts