Learn More
CD3e Armenian Hamster, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB2-100ug
 View more versions of this product
Product Code. 15931628
  • 131.25 € / 100µg
Specifications
CD3e
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd3e
Protein A/G Purified
RUO
12501
IgG
Flow Cytometry
145-2C11
Unconjugated
XP_006510029
Armenian Hamster
100 μg
Primary
Store at 2-4°C.