missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD4 Antibody (TRX1) - Humanized, Novus Biologicals™

Product Code. 30047379 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30047379 1 mg 1mg
30048168 100 μg 100µg
30045838 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30047379 Supplier Novus Biologicals™ Supplier No. NBP3283521mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone TRX1
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. P01730
Gene Alias CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Host Species Human
Immunogen CD4
Purification Method Protein A purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.