missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Rabbit, Clone: SPN/1766R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 6693-RBM6-P0
This item is not returnable.
View return policy
Specifications
| CD43 | |
| Cell surface | |
| SPN/1766R | |
| Unconjugated | |
| NP_003114 | |
| Rabbit | |
| 95, 115, or 135kDa | |
| Protein A/G Purified | |
| RUO | |
| 6693 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| SPN | |
| Recombinant full-length human SPN protein | |
| Paracortex in a tonsil or a reactive lymph node | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction