missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Rabbit, Clone: SPN/1766R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | CD43 |
|---|---|
| Clone | SPN/1766R |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15900908
|
EnquireBio
6693-RBM6-P0 |
20 μg | |||||||
|
15910908
|
EnquireBio
6693-RBM6-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD43 | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 6693 | |
| Recombinant full-length human SPN protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 95, 115, or 135kDa |
| SPN/1766R | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_003114 | |
| SPN | |
| IgG κ | |
| Protein A/G Purified | |
| Cell surface | |
| Paracortex in a tonsil or a reactive lymph node |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title