missing translation for 'onlineSavingsMsg'
Learn More

CD45.2 Mouse, APC-Cy7, Clone: 104, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB43-APC7-25ug

 View more versions of this product

Product Code. 15932028

  • 183.75 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD45.2
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
104
APC-Cyanine7
NP_001255215
Mouse
25 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.