missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.2 Mouse, APC-Cy7, Clone: 104, enQuireBio™
Mouse Monoclonal Antibody
183.75 € - 394.80 €
Specifications
| Antigen | CD45.2 |
|---|---|
| Clone | 104 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15932028
|
EnquireBio
QAB43-APC7-25ug |
25 μg |
183.75 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15922028
|
EnquireBio
QAB43-APC7-100ug |
100 μg |
394.80 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD45.2 | |
| 0.5mg/mL | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 19264 | |
| IgG2a κ | |
| Protein A/G Purified |
| 104 | |
| Flow Cytometry | |
| APC-Cyanine7 | |
| RUO | |
| NP_001255215 | |
| Ptprc | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title