Learn More
CD45RA Mouse anti-Rat, FITC, Clone: OX-33, BD
Mouse Monoclonal Antibody
159.43 € - 515.74 €
Specifications
| Antigen | CD45RA |
|---|---|
| Clone | OX-33 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811439
|
BD Biosciences
561886 |
50 μg |
166.00 € 159.43 € / Each Save 6.57 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15867898
|
BD Biosciences
554883 |
0.5 mg |
540.00 € 515.74 € / Each Save 24.26 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The OX-33 antibody reacts with a high-molecular-weight form of CD45 found only on B lymphocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
This antibody isroutinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD45RA | |
| 0.5mg/mL | |
| Monoclonal | |
| Mouse | |
| Rat | |
| Leukocyte common antigen purified from rat splenocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| OX-33 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.