missing translation for 'onlineSavingsMsg'
Learn More

CD61 Mouse anti-Human, PE, Clone: VI-PL2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561912

 View more versions of this product

Product Code. 15871459

  • ONLINE EXCLUSIVE PRICE
    62.10 €
    60.12 € / Each
    Save 1.98 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The VI-PL2 monoclonal antibody specifically binds to CD61. CD61 is a 105 kDa transmembrane glycoprotein that is also known as integrin β3 and platelet glycoprotein IIIa (GPIIIa or GP3A). It is expressed on platelets, megakaryocytes, osteoclasts and endothelia. Integrin β3 associates with gpIIa (CD41) to form the CD41/CD61 complex which mediates platelet adhesion and aggregation. CD61 also associates with CD51 to form the CD51/CD61 complex (vitronectin receptor). CD61 appears to bind to fibrinogen, fibronectin, vWF, vitronectin, and thrombospondin to mediate cell adhesion.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD61
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Baboon, Bovine, Cynomolgus, Human, Rhesus Monkey
IgG1 κ
Flow Cytometry
VI-PL2
Integrin β3; Integrin beta-3; GP3A; GPIIIa; ITGB3; ITB3
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.