Learn More
CD61 Mouse anti-Human, PE, Clone: VI-PL2, BD
Mouse Monoclonal Antibody
60.12 € - 239.00 €
Specifications
| Antigen | CD61 |
|---|---|
| Clone | VI-PL2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15871459
|
BD Biosciences
561912 |
25 Tests |
62.10 € 60.12 € / Each Save 1.98 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15818418
|
BD Biosciences
555754 |
100 Tests |
239.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The VI-PL2 monoclonal antibody specifically binds to CD61. CD61 is a 105 kDa transmembrane glycoprotein that is also known as integrin β3 and platelet glycoprotein IIIa (GPIIIa or GP3A). It is expressed on platelets, megakaryocytes, osteoclasts and endothelia. Integrin β3 associates with gpIIa (CD41) to form the CD41/CD61 complex which mediates platelet adhesion and aggregation. CD61 also associates with CD51 to form the CD51/CD61 complex (vitronectin receptor). CD61 appears to bind to fibrinogen, fibronectin, vWF, vitronectin, and thrombospondin to mediate cell adhesion.
Specifications
| CD61 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| VI-PL2 | |
| Monoclonal | |
| Mouse | |
| Baboon, Bovine, Cynomolgus, Human, Rhesus Monkey | |
| IgG1 κ | |
| Affinity Purified | |
| Integrin β3; Integrin beta-3; GP3A; GPIIIa; ITGB3; ITB3 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.