Learn More
CD62L / L-Selectin Rat, APC-Cy7, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB49-APC7-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15982108
  • 421.00 € / 100µg
Estimated Shipment: 03-07-2024
to see stock.
Specifications
CD62L / L-Selectin
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Sell
Protein A/G Purified
RUO
20343
IgG2a κ
Flow Cytometry
MEL-14
APC-Cyanine7
NP_001157531
Rat
100 μg
Primary
Store at 2-4°C.