missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L / L-Selectin Rat, APC-Cy7, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
155.00 € - 421.00 €
Specifications
Antigen | CD62L / L-Selectin |
---|---|
Clone | MEL-14 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15992108
|
EnquireBio
QAB49-APC7-25UG |
25 μg |
155.00 €
25µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982108
|
EnquireBio
QAB49-APC7-100UG |
100 μg |
421.00 €
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD62L / L-Selectin | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
20343 | |
IgG2a κ | |
Protein A/G Purified |
MEL-14 | |
Flow Cytometry | |
APC-Cyanine7 | |
RUO | |
NP_001157531 | |
Sell | |
Primary | |
Store at 2-4°C. |