missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Antibody (CD7/8496R), Novus Biologicals™
Rabbit Monoclonal Antibody
287.00 € - 519.00 €
Specifications
| Antigen | CD7 |
|---|---|
| Clone | CD7/8496R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690455
|
Novus Biologicals
NBP3-20469-100ug |
100 μg |
519.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655304
|
Novus Biologicals
NBP3-20469-20ug |
20 μg |
287.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| CD7 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Signal Transduction | |
| 10mM PBS with 0.05% BSA | |
| 924 | |
| IgG κ | |
| Protein A or G purified |
| CD7/8496R | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| Recombinant fragment (around aa 72-175) of human CD7 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title