missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79A Rabbit, anti-Human, Clone: 1F24, Proteintech
Rabbit Recombinant Monoclonal Antibody
149.00 € - 439.00 €
Specifications
| Antigen | CD79A |
|---|---|
| Clone | 1F24 |
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18047055
|
Proteintech
82747-4-RR-100UL |
100 μL |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040716
|
Proteintech
82747-4-RR-20UL |
20 μL |
149.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD79A antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CD79A. This antibody recognizes human antigen. The CD79A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CD79A | |
| 600 μg/mL | |
| Liquid | |
| Human | |
| CD79A, Ig alpha, IGA, MB 1, MB 1 membrane glycoprotein, MB1, Surface IgM associated protein | |
| Primary |
| 1F24 | |
| Western Blot, ELISA | |
| Rabbit | |
| 973 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title