missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (12.C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-64021-0.025MG
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Human, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CD8 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| CD8, CD8a molecule, LEU2, p32 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes the rabbit CD8 cell surface antigen, expressed by a subset of T lymphocytes with cytotoxic/suppressor activity. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry | |
| 12.C7 | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| CD8A | |
| Made to Rabbit CD8 | |
| 0.025 mg | |
| Innate Immunity, Stem Cell Markers | |
| 925 | |
| Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction