missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (12.C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
230.00 € - 526.00 €
Specifications
| Antigen | CD8 |
|---|---|
| Clone | 12.C7 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18086134
|
Novus Biologicals
NB100-64021-0.025MG |
0.025 mg |
230.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18734963
|
Novus Biologicals
NB100-64021 |
0.1 mg |
526.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal specifically detects CD8 in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| CD8 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Innate Immunity, Stem Cell Markers | |
| 925 | |
| Made to Rabbit CD8 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 12.C7 | |
| Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| CD8, CD8a molecule, LEU2, p32 | |
| CD8A | |
| IgG1 | |
| Protein G purified | |
| Recognizes the rabbit CD8 cell surface antigen, expressed by a subset of T lymphocytes with cytotoxic/suppressor activity. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title