Learn More
CD8 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566451
 View more versions of this product
Product Code. 15885059
  • ONLINE EXCLUSIVE PRICE
    206.00 €
    197.95 € / Each
    Save 8.05 €
    3% Off
Description
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8
Monoclonal
Brilliant Blue 700 (BB700)
25 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Flow Cytometry
RPA-T8
Mouse
RUO
Human, Baboon, Cynomolgus, Rhesus Macaque
Purified