missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
197.95 € - 413.02 €
Specifications
| Antigen | CD8 |
|---|---|
| Clone | RPA-T8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15885059
|
BD Biosciences
566451 |
25 Tests |
206.00 € 197.95 € / Each Save 8.05 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15895059
|
BD Biosciences
566452 |
100 Tests |
431.00 € 413.02 € / Each Save 17.98 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Specifications
| CD8 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human, Baboon, Cynomolgus, Rhesus Macaque | |
| Primary |
| RPA-T8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title