missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Antibody (101), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12771-100ul
This item is not returnable.
View return policy
Description
CD83 Monoclonal antibody specifically detects CD83 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD83 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 101 | |
| Flow Cytometry 1:25-1:100 | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 (Uniprot#: O88324; Met1-Arg133). | |
| 100 μg | |
| Dendritic Cell Markers | |
| 9308 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur