missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Antibody (101), Novus Biologicals™
Rabbit Monoclonal Antibody
298.00 € - 474.00 €
Specifications
| Antigen | CD83 |
|---|---|
| Clone | 101 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641477
|
Novus Biologicals
NBP3-12771-50ul |
50 μg |
298.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677668
|
Novus Biologicals
NBP3-12771-100ul |
100 μg |
474.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD83 Monoclonal antibody specifically detects CD83 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD83 | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 (Uniprot#: O88324; Met1-Arg133). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 101 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Dendritic Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 9308 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title