missing translation for 'onlineSavingsMsg'
Learn More

CDC42SE2 Antibody - BSA Free, Novus Biologicals™

Product Code. 30556628 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30556628 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30556628 Supplier Novus Biologicals Supplier No. NBP347663

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC42SE2 Polyclonal antibody specifically detects CDC42SE2 in Human samples. It is validated for ELISA,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42SE2
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:20-1:200
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias CDC42 small effector 2, FLJ21967, non-kinase Cdc42 effector protein SPEC2, Small effector of CDC42 protein 2, SPEC2CDC42 small effector protein 2
Host Species Rabbit
Immunogen CDC42 small effector 2. (Uniprot# Q9NRR3)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 56990
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.