missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDC45L Polyclonal antibody specifically detects CDC45L in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | CDC45L |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CDC45 cell division cycle 45-like (S. cerevisiae), cell division cycle 45 homolog (S. cerevisiae), cell division cycle 45-like 2, human CDC45, S.cerevisiae, homolog)-like |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CDC45L (NP_003495.1).,, Sequence:, MFVSDFRKEFYEVVQSQRVLLFVASDVDALCACKILQALFQCDHVQYTLVPVSGWQELETAFLEHKEQFHYFILINCGANVDLLDILQPDEDTIFFVCDTHRPVNVVNVYNDTQIKLLIKQDDDLEVPAYEDIFRDEEEDEEHSGNDSDGSEPSEKRTRLEEEIVEQTMRRRQRREWEARRRDILFDYEQYEYHGTSSAMVMFELAWMLSKDLNDMLWWAIVGLTDQWVQDKITQMKYVTDVGVLQRHVSRHNHRNEDEENTLSVDCTRI |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?