Learn More
CEBPB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23431-1-AP-150UL
This item is not returnable.
View return policy
Description
The CEBPB antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CEBPB. This antibody recognizes human, mouse antigen. The CEBPB antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| CEBPB | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| ChIP Assay, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| 150 μL | |
| 1051 | |
| IgG |