missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBPB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | CEBPB |
|---|---|
| Applications | ChIP Assay, Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16882345
|
Proteintech
23431-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16872345
|
Proteintech
23431-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CEBPB antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CEBPB. This antibody recognizes human, mouse antigen. The CEBPB antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| CEBPB | |
| Polyclonal | |
| Rabbit | |
| 1051 | |
| Primary |
| ChIP Assay, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title